Order Entry
Japan
ContactUsLinkComponent
LiChrosorb® RP-8, HPLC Columns
LiChrosorb® RP-8, HPLC Columns
Supplier:  HICHROM
   Sustainable Options Available
Certificates
undefined
LiChrosorb® RP-8, HPLC Columns
Supplier:  HICHROM
   Sustainable Options Available
Specifications
  • Catalog No:
  • LIRP8-5-10C5
  • LIRP8-10-250AF
  • LIRP8-5-250A
  • LIRP8-10-250A
  • LIRP8-5-125A
  • LIRP8-5-150A
  • LIRP8-10-125AF
  • LIRP8-10-200A
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Carbon load:
  • 9.5%
  • 9,5%
  • 9,5%
  • 9,5%
  • 9,5%
  • 9,5%
  • 9,5%
  • 9,5%
  • Endcapped:
  • No, No
  • No, No
  • No, No
  • No, No
  • No, No
  • No, No
  • No, No
  • No, No
  • Surface area:
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)