Order Entry
Japan
ContactUsLinkComponent
Polypropylene glycol, M.W. 2,000, Ultra Pure Grade
Polypropylene glycol, M.W. 2,000, Ultra Pure Grade
Catalog # E278-500G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Polypropylene glycol, M.W. 2,000, Ultra Pure Grade
Catalog # E278-500G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Acid Value:
    <1
  • APHA:
    ≤40
  • Moisture (KF):
    ≤0.15%
  • Viscosity (@ 100 °F):
    150 cS – 175 cS

Specification Test Results

About this item

 :
 :1025
Density:1.005 g/cm³ (20 °C)
Flash Pt:112.8 °C (closed cup)
Storage Temperature:Ambient
CAS Number:25322-69-4
Formula:H(OCH₂CH₂CH₂)nOH
MDL Number:MFCD00084446