Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR® Fluorescent DNA Quantitation Kit

VWR® Fluorescent DNA Quantitation Kit

Fluorescent DNA quantitation kit, 1 KIT
Catalog # 1B1558-KIT
Supplier:   VWR Chemicals
VWR® Fluorescent DNA Quantitation Kit
Catalog # 1B1558-KIT
Supplier:  VWR Chemicals
CAS Number:  
VWR® Fluorescent DNA Quantitation Kit
VWR® Fluorescent DNA Quantitation Kit

Fluorescent DNA quantitation kit, 1 KIT

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 KIT
Product Details & Documents

Fluorescent DNA Quantitation Kit uses fluorescent bisBenzimide H33258 dye to bind to AT sequences of the minor groove of double-stranded DNA. In the assay, the sample is excited at 360 nm and when the dye binds to dsDNA an emission spectrum at 460 nm is produced.

  • Highly specific quantitation of DNA
  • Sensitive to low nanogram per ml levels of DNA
  • Assay tolerates RNA and protein contaminants

Fluorescent DNA Quantitation Kit can be used to quantify dsDNA in the low nanogram per ml (10 ng/ml) range, up to 5 microgram per ml (5 µg/ml).

Delivery information: Kit includes 10X of Fluorescent DNA Assay Buffer (200 ml), Fluorescent DNA Dye (250 µl), and Fluorescent DNA Standard (1 ml).

Documents

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    Fluorescent DNA quantitation kit

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.