VWR®, Accessories for VisiScope® Epifluorescent Microscopes
Catalog # 630-2059
Supplier: VWR Collection
Specifications
- Description:Phase contrast set, complete, incl. IOS W-Plan objectives (10×, 20×, 40×, 100×) and BF/DF/PH condenser
- Seq:96
- Pk:1