Water, Ultrapure Bioreagent for molecular biology, sterile, J.T.Baker®
Catalog # 4221-02
Supplier: Avantor
CAS Number:
Specifications
- Pack type:PE bottle, narrow mouth
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:Passes Test
- DNase Activity (None Detected):Passes Test
- RNase Activity:Passes Test
- Protease Activity:Passes Test
Specification Test Results
About this item
:
:18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332