N-Methyl-2-pyrrolidone (NMP) ≥99% (by GC, corrected for water content), BAKER ANALYZED®, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:872-50-4
:
Formula:C₅H₉NO
MDL Number:MFCD00003193
:99.13 g/mol
Merck Index:13,06140
Boiling Pt:202 °C (1013 hPa)
Melting Pt:–24 °C
Density:1.033 g/cm³ (20 °C)
Flash Pt:91 °C
Storage Temperature:Ambient
EINECS:212-828-1
:GHS08
:GHS07
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99 %
- Assay (C₅H₉NO) (by GC, corrected for water):≥ 99 %
- Color (APHA):≤ 50
- Density (g/mL) at 25°C:1.024 - 1.028
- Water (H₂O)(by Karl Fischer titrn):≤ 0.20 %
Specification Test Results
Frequently Bought Together