Copper(II) sulphate pentahydrate 98.0-102.0% (by iodometry), fine crystals, AR® ACS, Macron Fine Chemicals™
4844-04
:
- Pack type:PE bottle, wide mouth
- Pk:500 g
- Seq:3645
- Meets ACS Reagent Chemical Requirements:98.0 - 102.0%
- For Laboratory,Research,or Manufacturing Use:≤0.001%
- ACS - Assay (CuSO₄·5H₂O) (by iodometry):98.0 - 102.0 %
- ACS - Appearance (blue, crystalline granules):Passes Test
- Solubility (5 in 50):Passes Test
- ACS - Chloride (Cl):≤0.001 %
- ACS - Insoluble Matter:≤0.005 %
- ACS - Iron (Fe):≤0.003 %
- ACS - Nitrogen Compounds (as N):≤0.002 %
- ACS - Calcium (Ca):≤0.005 %
- ACS - Nickel (Ni):≤0.005 %
- ACS - Potassium (K):≤0.01 %
- ACS - Sodium (Na):≤0.02 %
:
:249.69 g/mol
:13,02682
:2.284 g/cm³ (20 °C)
:Ambient
:231-847-6
:3288
:6.1,III
:01-2119520566-40
:GHS05
:GHS08
:GHS09
:7758-99-8
:CuSO₄·5H₂O
:MFCD00149681