

Bismuth(III) nitrate pentahydrate ≥98.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:10035-06-0
:
Formula:Bi(NO₃)₃·5H₂O
MDL Number:MFCD00149157
Boiling Pt:75…80 °C (1013 hPa)
Melting Pt:30 °C
Density:2.83 g/cm³
EINECS:233-791-8
UN:1477
ADR:5.1,II
:485.07 g/mol
:GHS03
:GHS05
Specification Test Results
- Meets ACS Reagent Chemical Requirements:≥98.0%
- For Laboratory,Research,or Manufacturing Use:≤0.005%
- ACS - Assay ((Bi(NO₃)₃·5H₂O)) (by EDTA titrn):≥98.0 %
- Insoluble in HNO₃:≤0.005 %
- Chloride (Cl):≤0.001 %
- Sulfate (SO₄):≤0.005 %
- Arsenic (As):≤0.001 %
- Calcium (Ca):≤0.005 %
- Copper (Cu):≤0.002 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Lead (Pb):≤0.002 %
- Potassium (K):≤0.01 %
- Silver (Ag):≤0.001 %
- Sodium (Na):≤0.02 %
Specification Test Results
Frequently Bought Together