Order Entry
Japan
ContactUsLinkComponent
Aluminium nitrate nonahydrate 98.0-102.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Oxidizing
Irritant
Aluminium nitrate nonahydrate 98.0-102.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0528-05
Supplier:  Avantor
CAS Number:  
undefined
Oxidizing
Irritant
Aluminium nitrate nonahydrate 98.0-102.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0528-05
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    98.0 - 102.0%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.005%
  • ACS - Assay (Al(NO₃)₃·9H₂O) (by EDTA titrn):
    98.0 - 102.0 %
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - pH of 5% Solution at 25°C:
    2.5 - 3.5
  • ACS - Calcium (Ca):
    ≤0.005 %
  • ACS - Chloride (Cl):
    ≤0.001 %
  • ACS - Magnesium (Mg):
    ≤0.001 %
  • ACS - Potassium (K):
    ≤0.002 %
  • ACS - Sodium (Na):
    ≤0.005 %
  • ACS - Sulfate (SO₄):
    ≤0.005 %
  • ACS - Heavy Metals (as Pb):
    ≤0.001 %
  • ACS - Iron (Fe):
    ≤0.002 %

Specification Test Results

About this item

 :
 :375.13 g/mol
Melting Pt:73 °C
Density:1.72 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:236-751-8
UN:1438
ADR:5.1,III
 :GHS03
 :GHS07
CAS Number:7784-27-2
Formula:Al(NO₃)₃·9H₂O
MDL Number:MFCD00149132