

Potassium hydroxide ≥85%, pellets, AR® ACS, Macron Fine Chemicals™
:1310-58-3
:
:KOH
:MFCD00003553
:56.11 g/mol
:12,07806
:1320 °C (1013 hPa)
:360 °C
:2.04 g/cm³ (20 °C)
:Ambient
:215-181-3
:1813
:8,II
:01-2119487136-33
:GHS05
:GHS07
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:= 86 %
- Assay (KOH):≥85 %
- Identification:Passes Test
- Calcium (Ca):≤0.005 %
- Chloride (Cl):≤0.01 %
- Heavy Metals (as Ag):≤0.001 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Nickel (Ni):≤0.001 %
- ACS - Magnesium (Mg):≤0.002 %
- Nitrogen Compounds (as N):≤0.001 %
- ACS - Phosphate (PO₄):≤5 ppm
- Sodium (Na):≤0.05 %
- Potassium Carbonate (K₂CO₃):≤2.0 %
- ACS - Sulfate (SO₄):≤0.003 %
Frequently Bought Together