



2,2,4-Trimethylpentane ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:540-84-1
:
Formula:(CH₃)₂CHCH₂C(CH₃)₃
MDL Number:MFCD00008943
:114.23 g/mol
Merck Index:13,05212
Boiling Pt:99.2 °C (1013 hPa)
Melting Pt:–107 °C
Density:0.692 g/cm³ (20 °C)
Flash Pt:–12 °C
EINECS:208-759-1
UN:1262
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Meets ACS Reagent Chemical Requirements:= 99.0 %
- For Laboratory,Research,or Manufacturing Use:= 10
- Assay ((CH₃)₃CCH₂CH(CH₃)₂) (by GC, corrected for water):≥ 99.0 %
- Color (APHA):≤ 10
- Residue after Evaporation:≤ 0.001 %
- Water-Soluble Titrable Acid, meq/g:≤ 0.0003
- Sulfur Compounds (as S):≤ 0.005 %
Specification Test Results
Frequently Bought Together