



n-Heptane ≥99.0% (by GC, corrected for water content), BAKER ANALYZED®, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:142-82-5
:
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544
:100.20 g/mol
Merck Index:13,04679
Boiling Pt:98.4 °C (1013 hPa)
Melting Pt:–90.6 °C
Density:0.6838 g/cm³ (20 °C)
Flash Pt:–4 °C
EINECS:205-563-8
UN:1206
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.0 %
- Assay (CH₃(CH₂)₅CH₃) (by GC, corrected for water):≥ 99.0 %
- Color (APHA):≤ 10
- Residue after Evaporation:≤ 0.001 %
Specification Test Results
Frequently Bought Together