Order Entry
Japan
ContactUsLinkComponent
Methyl ethyl ketone ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Flammable
Irritant
Methyl ethyl ketone ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 9319-22
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Irritant
Methyl ethyl ketone ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 9319-22
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Aluminium bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
    = 99.0 %
  • Assay (CH₃COCH₂CH₃) (by GC, corrected for water):
    ≥99.0 %
  • Color (APHA):
    ≤15
  • Residue after Evaporation:
    ≤0.001 %
  • Titrable Acid (µeq/g):
    ≤0.5
  • Water (by KF, volumetric):
    ≤0.05 %

Specification Test Results

About this item

 :
Merck Index:13,06097
Boiling Pt:79.6 °C (1013 hPa)
Melting Pt:–86 °C
Density:0.8054 g/cm³ (20 °C)
Flash Pt:–4 °C
ADR:3,II
 :Management of Change (MOC) category = N
 :72.11 g/mol
Storage Temperature:Ambient
EINECS:201-159-0
UN:1193
REACH:01-2119457290-43
 :GHS02
 :GHS07
CAS Number:78-93-3
Formula:C₂H₅COCH₃
MDL Number:MFCD00011648