Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, Tissue Processing/Embedding Cassettes, Premium

VWR®, Tissue Processing/Embedding Cassettes, Premium

Supplier:   VWR Collection
VWR®, Tissue Processing/Embedding Cassettes, Premium
Supplier:  VWR Collection
Certificates
VWR®, Tissue Processing/Embedding Cassettes, Premium
VWR®, Tissue Processing/Embedding Cassettes, Premium

2 Options Available Below

Product Details & Documents

These cassettes made of acetyl polymer feature a fluidic flow design for the optimisation of tissue processing and embedding.

  • Resistant to histological solvents
  • Chamber is 10% longer allowing for an increased specimen range
  • 45° angled print surface compatible with all automatic printing devices
  • Attached cover has a large labelling area and uses a frangible hinge
  • Unique design with vented side walls to maximise fluid exchange

The vented sides and rear wall design work exceptionally well with fatty, dense, and other difficult to process tissue samples.

Product Family Options

Product Information

  • Colour
    Availability
    Price
  • VWRU18000-004
    Tissue cassettes, Premium
    Tissue cassettes, Premium
    Turquoise
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - $620.72
    $620.72
    1 * 1.000 items
       $620.72

  • VWRU18000-006
    Tissue cassettes, Premium
    Tissue cassettes, Premium
    Green
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - $620.72
    $620.72
    1 * 1.000 items
       $620.72