Order Entry
Japan
ContactUsLinkComponent
VWR® Talon®, Multiple rod connector
VWR® Talon®, Multiple rod connector
Catalog # VWRU60079-053
Supplier:  VWR Collection
undefined
VWR® Talon®, Multiple rod connector
Catalog # VWRU60079-053
Supplier:  VWR Collection
Specifications
  • Pk:
    1
  • For rod Ø:
    13 mm
Specifications
About this item
Multiple rod connector holds lab-frame rods or extension clamps at 90° while allowing both horizontal and vertical adjustment of lattice after construction. Two adjacent rods fit into oval hole in connector that prevent turning when tightened.
  • Versatility and maximum adjustment capability at your fingertips
  • Create a variety of configurations via side and front holes
Ordering information: Two adjacent rods has to be ordered separately.