VWR® Talon®, Hosecock clamps, regular
Catalog # VWRU89202-614
Supplier: VWR Collection
Specifications
- Spanning width:17 mm
- Description:Hosecock clamp, stainless steel
- Seq:02
- Pk:1
Specifications
About this item
Hosecock clamps deliver precise flow control for tubing up to 17 mm diameter.
- Adjustment screw has an oversized head for easy one-hand operation and accurate regulation of fluid flow
- Tubing retainer screw on side of clamp keeps tubing from disengaging under varying fluid pressures, and can be raised to allow easy positioning of tubing
- Convex bearing surface reduces the amount of pressure required for complete closure; eliminates sharp edges that could cut into tubing