Order Entry
Japan
ContactUsLinkComponent
VWR®, Beta Radiation Boxes for Microcentrifuge Tube
VWR®, Beta Radiation Boxes for Microcentrifuge Tube
Catalog # VWRU32800-066
Supplier:  VWR Collection
undefined
VWR®, Beta Radiation Boxes for Microcentrifuge Tube
Catalog # VWRU32800-066
Supplier:  VWR Collection

Specifications

  • Description:
    Beta Radiation Storage Box
  • Dimensions:
    200×90×60 mm
  • Pk:
    1

Specifications

About this item

Acrylic, transparent.

  • Crushed ice or water fits under the rack
  • Holds 36 microcentrifuge tubes (1,5 ml)

Storage box with tight-fitting joints provides maximum protection from radioactive isotopes. Thickness: 9,5 mm.