Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, Desiccator Cabinets, Acrylic
VWR®, Desiccator Cabinets, Acrylic
Supplier:   VWR Collection
VWR®, Desiccator Cabinets, Acrylic
Supplier:  VWR Collection
Certificates
VWR®, Desiccator Cabinets, Acrylic
VWR®, Desiccator Cabinets, Acrylic
2 Options Available Below
Product Details & Documents
These acrylic desiccator cabinets can be used in refrigerators or at room temperature for dry storage of moisture-sensitive materials.
  • Fully removable, tray-like shelves have perforated bottoms and slide in/out on pre-set rails
  • Door opens from right to left and is secured with a stainless steel latch
Acrylic desiccator cabinets are fabricated from either clear or bronze acrylic, with a variable number of trays depending on model. Clear acrylic for easy viewing or bronze for light-sensitive materials. Bronze acrylic features stainless steel twist locking latches and hinges.
Product Family Options
Product Information
  • Shelves
    W×D×H ext.
    Availability
    Price
  • VWRU32800-012
    Desiccator cabinet, clear acrylic, 23,2 L
    Desiccator cabinet, clear acrylic, 23,2 L
    3
    305×305×305 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • VWRU32800-046
    Desiccator cabinet, bronze acrylic, 23,2 L
    Desiccator cabinet, bronze acrylic, 23,2 L
    3
    305×305×305 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items