Order Entry
Japan
ContactUsLinkComponent
VWR® Talon®, Swivel holder
VWR® Talon®, Swivel holder
Catalog # VWRU21572-603
Supplier:  VWR Collection
undefined
VWR® Talon®, Swivel holder
Catalog # VWRU21572-603
Supplier:  VWR Collection
Specifications
  • Spanning width:
    19 mm
  • Pk:
    1
Specifications
About this item
This Q-clamp post holder with center swivel capacity allows tilting of clamps at any angle in parallel planes. Made of nickel-plated zinc.
  • Allows tilting at any angle
  • Outside adjustment screw allows close proximity between objects being held