Order Entry
Japan
ContactUsLinkComponent
VWR® ColorFrost, Microscope Slides
VWR® ColorFrost, Microscope Slides
Catalog # VWRU48312-502
Supplier:  VWR Collection
undefined
VWR® ColorFrost, Microscope Slides
Catalog # VWRU48312-502
Supplier:  VWR Collection

Specifications

  • Colour:
    Green
  • Description:
    Microscope slides, ColorFrost
  • L×W:
    75×25 mm
  • Thickness:
    1 mm
  • Pk:
    72

Specifications

About this item

Slides frosted with a brilliantly coloured, opaque coating, which takes and retains most marking methods.

  • Easy to write on and to read: Key shows which side is up
  • No sticking or scratching, coating impervious to chemicals and reagents
  • Slides with raised printed writing surface that eliminates paper labels and re-labelling