Order Entry
Japan
ContactUsLinkComponent
VWR®, Microtube and Test Tube Racks, Floating
VWR®, Microtube and Test Tube Racks, Floating
Catalog # VWRU82017-630
Supplier:  VWR Collection
undefined
VWR®, Microtube and Test Tube Racks, Floating
Catalog # VWRU82017-630
Supplier:  VWR Collection
Specifications
  • For tubes:
    15,0 ml
  • Form:
    Parallelogram
  • No. of Holes:
    8
  • Colour:
    Blue
  • Pk:
    5
Specifications
About this item
Polyfoam, coloured.
  • These stands float and remain on the surface of the bath, preventing the vessels they contain from floating free
  • A variety of geometrical shapes enable individual and easily sortable arrangement in the water bath
  • With handles for easy insertion and removal
For incubating sample, centrifuge and microcentrifuge tubes in water baths.