Order Entry
Japan
ContactUsLinkComponent
Perchloric acid 70%, AR Select® ACS, Macron Fine Chemicals™
Oxidizing
Corrosive
Irritant
Perchloric acid 70%, AR Select® ACS, Macron Fine Chemicals™
Catalog # 8828-14
CAS Number:  
undefined
Oxidizing
Corrosive
Irritant
Perchloric acid 70%, AR Select® ACS, Macron Fine Chemicals™
Catalog # 8828-14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, clear, Mall
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    69.0 - 72.0 %
  • For Laboratory,Research,or Manufacturing Use:
    = 0.001 %
  • ACS - Assay (HClO₄):
    69.0 - 72.0 %
  • ACS - Chloride (Cl):
    ≤ 0.001 %
  • ACS - Color (APHA):
    ≤ 10
  • ACS - Heavy Metals (as Pb):
    ≤ 0.0001 %
  • ACS - Nitrogen Compounds (as N):
    ≤ 0.001 %
  • ACS - Residue after Ignition:
    ≤ 0.003 %
  • ACS - Silicate and Phosphate (as SiO₂):
    ≤ 0.0005 %
  • ACS - Sulfate (SO₄):
    ≤ 0.001 %
  • Determine by Flame Photometry & ICP - Aluminum (Al):
    ≤ 100 ppb
  • Determine by Flame Photometry & ICP - Barium (Ba):
    ≤ 100 ppb
  • Determine by Flame Photometry & ICP - Beryllium (Be):
    ≤ 10 ppb
  • Determine by Flame Photometry & ICP - Bismuth (Bi):
    ≤ 10 ppb
  • Determine by Flame Photometry & ICP - Boron (B):
    ≤ 1000 ppb
  • Determine by Flame Photometry & ICP - Cadmium (Cd):
    ≤ 50 ppb
  • Determine by Flame Photometry & ICP - Calcium (Ca):
    ≤ 1000 ppb
  • Determine by Flame Photometry & ICP - Chromium (Cr):
    ≤ 100 ppb
  • Determine by Flame Photometry & ICP - Cobalt (Co):
    ≤ 50 ppb
  • Determine by Flame Photometry & ICP - Copper (Cu):
    ≤ 100 ppb
  • Determine by Flame Photometry & ICP - Gallium (Ga):
    ≤ 500 ppb
  • Determine by Flame Photometry & ICP - Germanium (Ge):
    ≤ 500 ppb
  • Determine by Flame Photometry & ICP - Gold (Au):
    ≤ 500 ppb
  • Determine by Flame Photometry & ICP - Iron (Fe):
    ≤ 100 ppb
  • Determine by Flame Photometry & ICP - Lead (Pb):
    ≤ 100 ppb
  • Determine by Flame Photometry & ICP - Lithium (Li):
    ≤ 500 ppb
  • Determine by Flame Photometry & ICP - Magnesium (Mg):
    ≤ 100 ppb
  • Determine by Flame Photometry & ICP - Manganese (Mn):
    ≤ 50 ppb
  • Determine by Flame Photometry & ICP - Mercury (Hg):
    ≤ 100 ppb
  • Determine by Flame Photometry & ICP - Molybdenum (Mo):
    ≤ 10 ppb
  • Determine by Flame Photometry & ICP - Nickel (Ni):
    ≤ 100 ppb
  • Determine by Flame Photometry & ICP - Potassium (K):
    ≤ 500 ppb
  • Determine by Flame Photometry & ICP - Silicon (Si):
    ≤ 500 ppb
  • Determine by Flame Photometry & ICP - Silver (Ag):
    ≤ 100 ppb
  • Determine by Flame Photometry & ICP - Sodium (Na):
    ≤ 1000 ppb
  • Determine by Flame Photometry & ICP - Strontium (Sr):
    ≤ 100 ppb
  • Determine by Flame Photometry & ICP - Thallium (Tl):
    ≤ 50 ppb
  • Determine by Flame Photometry & ICP - Tin (Sn):
    ≤ 500 ppb
  • Determine by Flame Photometry & ICP - Vanadium (V):
    ≤ 10 ppb
  • Determine by Flame Photometry & ICP - Zinc (Zn):
    ≤ 100 ppb
  • Determine by Flame Photometry & ICP - Zirconium (Zr):
    ≤ 10 ppb

Specification Test Results

About this item

 :
 :100.46 g/mol
Boiling Pt:160…181 °C (1013 hPa)
Melting Pt:–112.2 °C
Density:1.53…1.768 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-512-4
UN:1873
ADR:5.1,I
 :Management of Change (MOC) category = N
 :GHS03
 :GHS05
 :GHS07
CAS Number:7601-90-3
Formula:HClO₄
MDL Number:MFCD00011325