Order Entry
Japan
ContactUsLinkComponent
 
Sodium hydroxide 0.02 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
 :  Avantor
 
undefined
Sodium hydroxide 0.02 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
 :  Avantor
 

 

  • For Laboratory,Research,or Manufacturing Use:
    0.0195 - 0.0205
  • Protect from air to avoid absorption of carbon dioxide.:
    = 1 ppm
  • Normality:
    0.0195 - 0.0205
  • Chloride (Cl):
    ≤1 ppm
  • Trace Impurities - Heavy Metals:
    ≤0.5 ppm
  • Trace Impurities - Iron (Fe):
    ≤0.5 ppm

 

 

Frequently Bought Together