Order Entry
Japan
ContactUsLinkComponent
VWR®, Waste Container, Heavy Duty, Floor Model
VWR®, Waste Container, Heavy Duty, Floor Model
Catalog # VWRU89030-154
Supplier:  VWR Collection
undefined
VWR®, Waste Container, Heavy Duty, Floor Model
Catalog # VWRU89030-154
Supplier:  VWR Collection
Specifications
  • W×D×H:
    673×330×330 mm
  • Pk:
    1
Specifications
About this item
Acrylic, clear.
  • Available with wheels for easy mobility
  • Clear acrylic casing permits visibility of warning insignias or biohazard bags
  • With hinged lid for convenient placement of waste materials
These waste containers are ideal for keeping sharp objects and liquid waste contained in the event of waste box leakage.