るつぼ、磁器
Supplier: VWR Collection
Certificates
About this item
Porcelain, glased inside and out, except base and rim.
- Withstand maximum temperature of 1150 °C
- Not recommended for minerals or metals
Lids are available separately.
Lids for crucibles
Product Information
Frequently Bought Together