バッグ、バイオハザード
These bags provide an efficient, safe solution for the messy and potentially dangerous process of disposing of biohazardous waste material.
- PP, 38 µm thick
- Perfect adaptability to standard laboratory disposal routines, including steam autoclaving
- With biohazard warning and printed area
- For use at 135 °C
: Twist ties are included.
Frequently Bought Together