VWR®, Staining Jars according to Hellendahl
Catalog # VWRU25460-907
Supplier: VWR Collection
Specifications
- Description:Staining jar acc. to Hellendhal, for 16 slides, with cover
- W×D×H:58×54×86 mm
- Pk:1
Specifications
About this item
PMP, with cover.
Holds 76×26 mm slides vertically. Eight grooves accommodate 16 slides back to back. Xylene can cause etching after 2 days at room temperature.