VWR® Tri-Pour®, Beakers, disposable, PP
VWRU25384-160
: 25384-160
PP, graduated.
- With three pouring lips for dripless dispensing
- Graduatations with accuracy of ±5%
- Write-on cardboard caps ensure spillproof, leak-resistant closure
These beakers may also be used for measuring. Resistant to strong acids and bases, organic solvents, and commonly used laboratory reagents.
Frequently Bought Together