VWR® Durac™, Baumé hydrometer
Supplier: VWR Collection
Certificates
About this item
Baumé hydrometer designed for determining the specific gravity and Baumé in liquids lighter then water.
- Dual scale
- Individually serialised
- Lead free metal ballast
Specially used in chemistry, pharmacology, food and beverage, and in the starch industry.
Certifications: NIST and DAkkS. Calibration conforms to A2LA accreditions.
Delivery: Supplied with a multi-language statement of accuracy.
Specifications
- Catalog No:
- VWRU34670-161
- VWRU34670-140
- Reference temperature:
- 15,6 °C