Order Entry
Japan
ContactUsLinkComponent
 
VWR® Silhouette, Glove Dispenser
Catalog # VWRU10031-908
Supplier:  VWR Collection
undefined
VWR® Silhouette, Glove Dispenser
Catalog # VWRU10031-908
Supplier:  VWR Collection

Specifications

  • Description:
    Glove dispenser for slim gloves boxes, Silhouette
  • W×D×H:
    258,7×79,4×536,5 mm
  • Pk:
    1

Specifications

About this item

This glove boxes dispenser is designed to fit for slim profile glove boxes.

  • Fits most manufacturers slim boxes
  • Glove boxes of up to 70 mm depth