Order Entry
Japan
ContactUsLinkComponent
VWR®, Nitrile gloves
VWR®, Nitrile gloves
Supplier:  VWR Collection
Certificates
VWR®, Nitrile gloves
Supplier:  VWR Collection
Specifications
  • Catalog No:
  • 82026-430
  • 82026-424
  • 82026-428
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Finish:
  • Textured
  • Textured
  • Textured
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,1 mm
  • 0,1 mm
  • 0,1 mm
  • Finger thickness:
  • 0,14 mm
  • 0,14 mm
  • 0,14 mm
  • Palm thickness:
  • 0,12 mm
  • 0,12 mm
  • 0,12 mm