VWR®, Overalls, Gamma Irratiated
Supplier: VWR Collection
Certificates
About this item
These gamma irradiated overalls are manufactured from a specially formulated breathable microporous fabric that provides significant fluid and barrier protection. Manufactured and packaged in an ISO Class 8 (Class 100 000) cleanroom.
- Gamma irradiated to a Sterility Assurance Level (SAL) of 10⁻⁴
- Excellent fluid and particulate barrier
- Extremely low level of particle shedding
- Superior breathability and water vapor transmission rate
- Soft, cloth-like fabric
Tested and manufactured in an ISO certified facility under stringent process controls to ensure that each product meets exacting quality standards and performs to specification. Products are validated through independent lab testing.
Colour: White
Specifications
- Catalog No:
- 414004-445
- 414004-446
- 414004-444
- 414004-454
- VWRU89136-080
- VWRU89136-082
- 414004-452
- 414004-443
- 414004-453
- 414004-449
- 414004-451
- 414004-447
- 414004-448
- VWRU89136-084
- VWRU89136-086
- VWRU414004-450
- Garment application:
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory