

1-Pentanol mixture of isomers ≥98.0%, AR®, Macron Fine Chemicals™
Catalog # MACR2996-08
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 98.0 %
- Assay (CH₃(CH₂)₃CH₂OH) (by GC):≥98.0 %
- Residue after Evaporation:≤0.003 %
Specification Test Results
About this item
:
:88.15 g/mol
Merck Index:13,07195
Boiling Pt:137.9 °C (1013 hPa)
Melting Pt:–79 °C
Density:0.81 g/cm³ (20 °C)
Flash Pt:48 °C
Storage Temperature:Ambient
EINECS:200-752-1
UN:1105
ADR:3,III
:GHS02
:GHS07
CAS Number:71-41-0
Formula:H₃C(CH₂)₃CH₂OH
MDL Number:MFCD00002977