



Mercury(II) iodide ≥99.0%, red powder, BAKER ANALYZED® ACS, J.T.Baker®
2610-01
: Avantor
:
- Pack type:Poly bottle, white, blue T/E cap
- Pk:500 g
- Seq:3645
- Meets ACS Reagent Chemical Requirements:≥99.0%
- For Laboratory,Research,or Manufacturing Use:≤0.05%
- ACS - Assay (HgI₂):≥99.0 %
- ACS - Solubility in Potassium Iodide Solution:Passes Test
- ACS - Mercurous Mercury (as Hg):≤0.1 %
- ACS - Soluble Mercury Salts (as Hg):≤0.05 %
:
:454.4 g/mol
:13,05905
:354 °C (1013 hPa)
:259 °C
:6.36 g/cm³ (20 °C)
:Ambient
:231-873-8
:1638
:6.1,II
:GHS06
:GHS08
:GHS09
:GHS07
:7774-29-0
:HgI₂
:MFCD00011044