Order Entry
Japan
ContactUsLinkComponent
 
Lead(II) oxide (Litharge) ≥99%, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 2338-01
Supplier:  Avantor
CAS Number:  
undefined
Lead(II) oxide (Litharge) ≥99%, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 2338-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 99.0 %
  • ACS - Assay (PbO):
    ≥ 99.0 %
  • ACS - Chloride (Cl):
    ≤ 0.002 %
  • ACS - Nitrate (NO₃):
    ≤ 0.01 %
  • ACS - Calcium (Ca):
    ≤ 0.005 %
  • ACS - Copper (Cu):
    ≤ 0.005 %
  • ACS - Potassium (K):
    ≤ 0.005 %
  • ACS - Silicon (Si), For Information Only:
    = 0.02 %
  • ACS - Sodium (Na):
    ≤ 0.02 %
  • Trace Impurities - Iron (Fe):
    ≤ 5 ppm
  • Trace Impurities - Silver (Ag):
    ≤ 5.000 ppm
  • Average Particle Diameter, µm (APD), For Information Only:
    For Information Only
  • Specific Surface Area, m/g, For Information Only:
    For Information Only
  • Bulk Density (g/cc), For Information Only:
    For Information Only

Specification Test Results

About this item

 :
Merck Index:13,05431
Boiling Pt:1470 °C (1013 hPa)
Melting Pt:888 °C
Density:9.5 g/cm³ (20 °C)
EINECS:215-267-0
UN:2291
ADR:6.1,III
 :223.21 g/mol
 :GHS08
 :GHS07
 :GHS09
CAS Number:1317-36-8
Formula:PbO
MDL Number:MFCD00011164