Hydrogen peroxide solution 30%, AR Select® ACS for trace metal analysis, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:7722-84-1
:
Formula:H₂O₂
MDL Number:MFCD00011333
Boiling Pt:∼107 °C (1013 hPa)
Melting Pt:< 0 °C
Density:1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-765-0
UN:2014
ADR:5.1,II
:34.01 g/mol
:GHS03
:GHS05
:GHS06
:GHS08
Specification Test Results
- Meets ACS Reagent Chemical Requirements:30.0 - 32.0 %
- For Laboratory,Research,or Manufacturing Use:= 10
- ACS - Assay (H₂O₂):30.0 - 32.0 %
- ACS - Color (APHA):≤10
- ACS - Residue after Evaporation:≤20 ppm
- ACS - Free Acid (µeq/g):≤0.6
- Trace Impurities - Aluminum (Al):≤0.200 ppm
- Ammonium (NH₄):≤5 ppm
- Arsenic and Antimony (as As):≤0.010 ppm
- Trace Impurities - Barium (Ba):≤0.010 ppm
- Trace Impurities - Beryllium (Be):≤0.010 ppm
- Trace Impurities - Bismuth (Bi):≤0.010 ppm
- Trace Impurities - Boron (B):≤0.010 ppm
- Trace Impurities - Cadmium (Cd):≤0.010 ppm
- Trace Impurities - Calcium (Ca):≤0.100 ppm
- Chloride (Cl):≤2 ppm
- Trace Impurities - Chromium (Cr):≤0.040 ppm
- Trace Impurities - Cobalt (Co):≤0.010 ppm
- Trace Impurities - Copper (Cu):≤0.010 ppm
- Trace Impurities - Gallium (Ga):≤0.010 ppm
- Trace Impurities - Germanium (Ge):≤0.020 ppm
- Trace Impurities - Gold (Au):≤0.010 ppm
- ACS - Heavy Metals (as Pb):≤1 ppm
- Trace Impurities - Iron (Fe):≤0.050 ppm
- Trace Impurities - Lead (Pb):≤0.010 ppm
- Trace Impurities - Lithium (Li):≤0.010 ppm
- Trace Impurities - Magnesium (Mg):≤0.010 ppm
- Trace Impurities - Manganese (Mn):≤0.050 ppm
- Trace Impurities - Mercury (Hg):≤0.001 ppm
- Trace Impurities - Molybdenum (Mo):≤0.010 ppm
- Trace Impurities - Nickel (Ni):≤0.020 ppm
- Nitrate (NO₃):≤2 ppm
- Phosphate (PO₄):≤2 ppm
- Trace Impurities - Potassium (K):≤0.400 ppm
- Trace Impurities - Silver (Ag):≤0.050 ppm
- Trace Impurities - Sodium (Na):≤0.050 ppm
- Trace Impurities - Strontium (Sr):≤0.010 ppm
- ACS - Sulfate (SO₄):≤5 ppm
- Trace Impurities - Thallium (Tl):≤0.050 ppm
- Trace Impurities - Tin (Sn):≤0.500 ppm
- Trace Impurities - Titanium (Ti):≤0.010 ppm
- Trace Impurities - Vanadium (V):≤0.010 ppm
- Trace Impurities - Zinc (Zn):≤0.100 ppm
- Trace Impurities - Zirconium (Zr):≤0.010 ppm
Specification Test Results
Frequently Bought Together