Order Entry
Japan
ContactUsLinkComponent
 
 
 
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
 
undefined
 
 
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
 

 

  • Assay:
    Min. 99.0 %
  • IR Spectrum:
    Passes test
  • Colouration:
    Max. 10 APHA
  • Evaporation residue:
    Max. 10 ppm
  • Peroxides (as H2O2):
    Max. 1 ppm
  • Water:
    Max. 0.05 %

 

Frequently Bought Together