Order Entry
Japan
ContactUsLinkComponent
 
Titration solvent ASTM D-5776, VOLUSOL®
Catalog # 5449.2500
Supplier:  VWR Chemicals
CAS Number:  
undefined
Titration solvent ASTM D-5776, VOLUSOL®
Catalog # 5449.2500
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Density (20/4):
    1,020 − 1,060
  • Control:
    Passes test

Specification Test Results

About this item

 :
UN:2790
ADR:8,II
 :GHS02
 :GHS05
 :GHS08
 :GHS07
Storage Temperature:Ambient