VWR®, Bovine serum albumin (BSA)
K720-50ML
:
- Pack type:Plastic bottle
- Pk:50 mL
- Source:Serum
- Species:Bovine
- Protein synonyms:BSA|Fraction V|Cohn Fraction V
- Protein/peptide name:Bovine Serum Albumin
- Grade:for biotechnology
Concentrated 20% BSA solution (Albumin, Bovine, Fraction V – Cold Alcohol Isolation) in 0,85% sodium chloride and contains 0,1% sodium azide (as a preservative for stability).