$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Japan
ContactUsLinkComponent
Tubing
Transfer Tubing
VWR®, Accessories for VWR® Refrigerated and Heated Circulator Tubings
VWR®, Accessories for VWR® Refrigerated and Heated Circulator Tubings
Catalog # 462-0305
Supplier:
VWR Collection
Expand image
VWR®, Accessories for VWR® Refrigerated and Heated Circulator Tubings
Catalog # 462-0305
Supplier:
VWR Collection
Specifications
Description:
Tube Clamp for 1/4" (6,3 mm) and 3/8" (9,5 mm) tubing, stainless steel
Seq:
03
Pk:
1
Specifications
About this item
Product Details
Product Details