Order Entry
Japan
ContactUsLinkComponent
Silver nitrate 0.025 mol/l (0.025 N)
Flammable
Irritant
Silver nitrate 0.025 mol/l (0.025 N)
Catalog # 88017.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Irritant
Silver nitrate 0.025 mol/l (0.025 N)
Catalog # 88017.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
 :169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
UN:1993
ADR:3,II
 :GHS02
 :GHS07
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414