
Phenolphthalein, powder, BAKER ANALYZED® ACS suitable as pH-indicator, J.T.Baker®
Catalog # 2870-01
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:Passes Test
- ACS - Clarity of Solution:Passes Test
- Visual Transition Interval - pH…8.0 (Colorless):Passes Test
- Visual Transition Interval - pH…10.0 (Red):Passes Test
Specification Test Results
About this item
:
:318.33 g/mol
Merck Index:14,07243
Melting Pt:258…262 °C
Density:1.3 g/cm³ (20 °C)
EINECS:201-004-7
REACH:01-2119498295-24
:GHS08
Storage Temperature:Ambient
CAS Number:77-09-8
Formula:C₂₀H₁₄O₄
MDL Number:MFCD00005913