Order Entry
Japan
ContactUsLinkComponent
Mercury (II) potassium iodide solution Reag. Ph. Eur. 1071500
Toxic
Environment
Mercury (II) potassium iodide solution Reag. Ph. Eur. 1071500
Catalog # 87921.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Environment
Mercury (II) potassium iodide solution Reag. Ph. Eur. 1071500
Catalog # 87921.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :786.4 g/mol
UN:2024
ADR:6.1,III
 :GHS06
 :GHS09
CAS Number:7783-33-7
Formula:HgI₄K₂
MDL Number:MFCD00049656