



Mercuric chloride solution Reag. Ph. Eur. 1052201
Catalog # 87865.290
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
UN:1624
ADR:6.1,II
:GHS06
:GHS05
:GHS08
:GHS09