VWR® Q Path®, Biopsy Pads
Supplier: VWR Collection
Jump to:
About this item
Polyester urethane foam. To hold small biopsies in place and prevent them from being lost. Enables fluids to circulate freely during processing.
- Individually cut
- Ready to use
- Solvent resistant
Packed in bags of 500 pieces.
2 Options Available Below
Product Details & Documents
Polyester urethane foam. To hold small biopsies in place and prevent them from being lost. Enables fluids to circulate freely during processing.
- Individually cut
- Ready to use
- Solvent resistant
Packed in bags of 500 pieces.