Order Entry
Japan
ContactUsLinkComponent
VWR® TBS/Tween® 20X Washing Buffer, Liquid Concentrate, UltraPure Grade
VWR® TBS/Tween® 20X Washing Buffer, Liquid Concentrate, UltraPure Grade
Catalog # K873-1L
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® TBS/Tween® 20X Washing Buffer, Liquid Concentrate, UltraPure Grade
Catalog # K873-1L
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pk:
    1 L
  • Description:
    TBS/Tween® washing buffer, 20X liquid concentrate, Ultra Pure Grade
Specifications
About this item
Convenient liquid concentrate that produces 10 litres of 1X buffer for blotting and ELISA applications.
  • Prevents high background by effectively removing unbound reagents