Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR® Blocking buffer, fish gelatin, 10% solution

VWR® Blocking buffer, fish gelatin, 10% solution

Fish gelatin, 10% solution, blocking buffer, Plastic bottle, 500 mL
Catalog # M319-500ML
Supplier:   VWR Chemicals
VWR® Blocking buffer, fish gelatin, 10% solution
Catalog # M319-500ML
Supplier:  VWR Chemicals
CAS Number:  
VWR® Blocking buffer, fish gelatin, 10% solution
VWR® Blocking buffer, fish gelatin, 10% solution

Fish gelatin, 10% solution, blocking buffer, Plastic bottle, 500 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 500 mL
Product Details & Documents

A non mammalian blocking solution that will not cross-react with mammalian antibodies.

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    Fish gelatin, 10% solution, blocking buffer

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.