Order Entry
Japan
ContactUsLinkComponent
 
 
 
 
Alcohol, anhydrous 94.0-96.0% (v/v), PHOTREX® for spectrophotometry, for liquid chromatography (denatured with 5% isopropanol and 5% methanol), J.T.Baker®
 :  Avantor
 
undefined
 
 
 
Alcohol, anhydrous 94.0-96.0% (v/v), PHOTREX® for spectrophotometry, for liquid chromatography (denatured with 5% isopropanol and 5% methanol), J.T.Baker®
 :  Avantor
 

 

  • For Laboratory,Research,or Manufacturing Use:
    94.0 - 96.0 %
  • Assay Specially Denatured Alcohol Formula 3A (200 proof)(v/v):
    94.0 - 96.0 %
  • Isopropyl Alcohol (2-propanol)(v/v):
    4.0 - 6.0 %
  • Ethanol (v/v):
    88.5 - 92.5 %
  • Methanol (v/v):
    3.5 - 5.5 %
  • Appearance:
    Passes Test
  • Color (APHA):
    ≤5
  • Residue after Evaporation:
    ≤5 ppm
  • Specific Gravity at 25°C:
    0.785 - 0.795
  • Water (H₂O):
    ≤0.1 %
  • Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 210 nm:
    ≤1.00
  • Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 220 nm:
    ≤0.50
  • Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 230 nm:
    ≤0.20
  • Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 250 nm:
    ≤0.05
  • Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 270 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 400 nm:
    ≤0.01
  • Substances Reducing Permanganate:
    Passes Test

 

 

Frequently Bought Together