Order Entry
Japan
ContactUsLinkComponent
Iron, powder, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Flammable
Iron, powder, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 2226-01
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Iron, powder, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 2226-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 96.0 %
  • Assay (Fe):
    ≥96.0 %
  • Insoluble in H₂SO₄:
    ≤0.2 %
  • Water-Soluble Substances:
    ≤0.03 %
  • Sulfide (S):
    ≤0.03 %
  • Lead (Pb):
    ≤0.005 %
  • Trace Impurities - Arsenic (As):
    ≤5.000 ppm

Specification Test Results

About this item

 :
 :55.85 g/mol
Boiling Pt:3000 °C (1013 hPa)
Melting Pt:1536 °C
Density:7.86 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-096-4
UN:3089
ADR:4.1,III
 :GHS02
CAS Number:7439-89-6
Formula:Fe
MDL Number:MFCD00010999