Order Entry
Japan
ContactUsLinkComponent
 
Irritant
Environment
Copper(II) sulphate, anhydrous ≥98.5% (by iodometry), powder, BAKER ANALYZED®, J.T.Baker®
Catalog # 1850-01
Supplier:  Avantor
CAS Number:  
undefined
Irritant
Environment
Copper(II) sulphate, anhydrous ≥98.5% (by iodometry), powder, BAKER ANALYZED®, J.T.Baker®
Catalog # 1850-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 98.5 %
  • Assay (CuSO₄) (by iodometry):
    ≥98.5 %
  • Loss on Drying:
    ≤1.0 %
  • Calcium (Ca):
    ≤0.005 %
  • Iron (Fe):
    ≤0.005 %
  • Nickel (Ni):
    ≤0.005 %
  • Potassium (K):
    ≤0.01 %
  • Sodium (Na):
    ≤0.02 %
  • Lead (Pb):
    ≤2.0 ppm

Specification Test Results

About this item

 :
 :159.61 g/mol
Merck Index:12,02722
Melting Pt:200 °C
Density:3.603 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-847-6
UN:3288
ADR:6.1,III
 :GHS07
 :GHS09
CAS Number:7758-98-7
Formula:CuSO₄
MDL Number:MFCD00010981